1-Bromo-4-chloro-2-fluorobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19570-06
A19570-14
CAAAA19570-06EA
35.08
CAD
CAAAA19570-06
CAAAA19570-14
1-Bromo-4-chloro-2-fluorobenzene 98%
1-Bromo-4-chloro-2-fluorobenzene
Purity: 98%
CAS No.: 1996-29-8
Molecular Formula: C6H3BrClF
Formula Weight: 209.45
Flash Point: 92°(197°F)
Refractive Index: 1.5540
Boiling Point: 70-72°/11mm
Beilstein Reference: 2081083
MDL No.: MFCD00079708
Formula:
C₆H₃BrClF Boiling Pt: 70…72 °C (11 mmHg) Density: 1,678 Flash Pt: 92 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00079708 CAS Number: 1996-29-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...