3-Methylbenzophenone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: m-Methylbenzophenone

A19568-09 A19568-18
CAAAA19568-09EA 84.69 CAD
CAAAA19568-09 CAAAA19568-18
3-Methylbenzophenone 99%
3-Methylbenzophenone

Purity: 99% CAS No.: 643-65-2 Molecular Formula: C14H12O Formula Weight: 196.25 Flash Point: 110°(230°F) Refractive Index: 1.5980 Boiling Point: 316° Beilstein Reference: 2045488 MDL No.: MFCD00008535

Formula: C₁₄H₁₂O
Boiling Pt: 316 °C
Melting Pt: 0…2 °C
Density: 1.093
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008535
CAS Number: 643-65-2
EINECS: 211-401-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR