Thiamine hydrochloride, (max. 5% H₂O) 99% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

A19560-0B A19560-22 A19560-30
CAAAAA19560-0BEA 690.46 CAD
CAAAAA19560-0B CAAAAA19560-22 CAAAA19560-30
Thiamine hydrochloride, (max. 5% H₂O) 99% (dry weight)
Thiamine hydrochloride

Purity: 99% CAS No.: 67-03-8 Molecular Formula: C12H17ClN4OS.HCl Formula Weight: 337.28 Physical Form: Cryst powder Sensitivity: Light Sensitive and Hygroscopic Appearance: White to pale yellow Melting Point: ca 250° dec. Beilstein Reference: 3851771 Merck Reference: 14,9295 MDL No.: MFCD00012780

Formula: C₁₂H₁₈Cl₂N₄OS
MW: 337.27 g/mol
Melting Pt: ∼ 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012780
CAS Number: 67-03-8
EINECS: 200-641-8
Merck Index: 14,9295

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR