2-Amino-p-cresol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-4-hydroxytoluene, 1-Amino-2-hydroxy-5-methylbenzene, 2-Amino-4-methylphenol, 2-Hydroxy-5-methylaniline

A19546-18 A19546-30
CAAAA19546-18EA 66.36 CAD
CAAAA19546-18 CAAAA19546-30
2-Amino-p-cresol 98%
2-Amino-p-cresol

Purity: 98% CAS No.: 95-84-1 Molecular Formula: C7H9NO Formula Weight: 123.16 Melting Point: 134-138° Beilstein Reference: 606494

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 259
Melting Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00007699
CAS Number: 95-84-1
EINECS: 202-457-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR