1-Chloro-3,4-dibromo-5-fluorobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19536-06
A19536-14
CAAAA19536-06EA
94.64
CAD
CAAAA19536-06
CAAAA19536-14
1-Chloro-3,4-dibromo-5-fluorobenzene 98%
1-Chloro-3,4-dibromo-5-fluorobenzene
Purity: 98%
CAS No.: 208186-78-1
Molecular Formula: C6H2Br2ClF
Formula Weight: 288.35
Melting Point: 37-39°
MDL No.: MFCD00143441
Formula:
C₆H₂Br₂ClF Boiling Pt: 242 °C (1013 hPa) Melting Pt: 37…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143441 CAS Number: 208186-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...