2-Acetyl-1-naphthol 99%

Supplier: Thermo Scientific Chemicals
Warning

A19532-14 A19532-22
CAAAA19532-14EA 90.22 CAD
CAAAA19532-14 CAAAA19532-22
2-Acetyl-1-naphthol 99%
2-Acetyl-1-naphthol

Purity: 99% CAS No.: 711-79-5 Molecular Formula: C12H10O2 Formula Weight: 186.21 Melting Point: 98-100° Beilstein Reference: 1868084

Formula: C₁₂H₁₀O₂
Boiling Pt: 110 - 115 °C (0.5 -rr)
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003963
CAS Number: 711-79-5
EINECS: 211-918-8
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR