Bromocresol purple

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 5',5''-Dibromo-o-cresolsulfonphthalein

A19530-09 A19530-18
CAAAA19530-09EA 59.95 CAD
CAAAA19530-09 CAAAA19530-18
Bromocresol purple
Bromocresol purple

CAS No.: 115-40-2 Molecular Formula: C21H16Br2O5S Formula Weight: 540.24 Melting Point: ca 240° dec. Beilstein Reference: 359618 Merck Reference: 14,1387 MDL No.: MFCD00011681

Formula: C₂₁H₁₆Br₂O₅S
MW: 540.23 g/mol
Melting Pt: 242 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011681
CAS Number: 115-40-2
EINECS: 204-087-8
Merck Index: 14,1387

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR