2-Nitroanisole 99%

Supplier: Thermo Scientific Chemicals
Danger

A19518-0E A19518-36
CAAAA19518-0EEA 699.94 CAD
CAAAA19518-0E CAAAA19518-36
2-Nitroanisole 99%
2-Nitroanisole

Purity: 99% CAS No.: 91-23-6 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 142°(287°F) Refractive Index: 1.5610 Melting Point: 9-10° Boiling Point: 272-273° Beilstein Reference: 1868032 Merck Reference: 14,6585 UN No.: 2730 MDL No.: MFCD00007096

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 272…273 °C
Melting Pt: 9…10 °C
Density: 1.254
Flash Pt: 142 °C (287 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007096
CAS Number: 91-23-6
EINECS: 202-052-1
UN: 2730
ADR: 6.1,III
Merck Index: 14,6585

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR