Ethyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A19492-14
A19492-22
A19492-36
CAAAA19492-14EA
102.54
CAD
CAAAA19492-14
CAAAA19492-22
CAAAA19492-36
Ethyl isothiocyanate 97%
Ethyl isothiocyanate
Purity: 97%
CAS No.: 542-85-8
Molecular Formula: CH3CH2NCS
Formula Weight: 87.14
Flash Point: 32°(89°F)
Refractive Index: 1.5110
Odor: Pungent
Melting Point: -6°
Boiling Point: 130-132°
Beilstein Reference: 1737705
Merck Reference: 14,3815
UN No.: 2929
MDL No.: MFCD00004820
Formula:
C₃H₅NS Boiling Pt: 130…132 °C Melting Pt: –6 °C Density: 0.995 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004820 CAS Number: 542-85-8 EINECS: 208-831-2 UN: 3286 ADR: 3,II Merck Index: 14,3815 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...