Decanoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A19486-22
A19486-36
CAAAA19486-22EA
61.15
CAD
CAAAA19486-22
CAAAA19486-36
Decanoyl chloride 98%
Decanoyl chloride
Purity: 98%
CAS No.: 112-13-0
Molecular Formula: CH3(CH2)8COCl
Formula Weight: 190.71
Flash Point: 106°(223°F)
Refractive Index: 1.4410
Odor: Irritating
Melting Point: -35°
Boiling Point: 232°
Beilstein Reference: 507055
UN No.: 3265
MDL No.: MFCD00000771
Formula:
C₁₀H₁₉ClO Boiling Pt: 232 °C Melting Pt: –35 °C Density: 0.925 Flash Pt: 106 °C (223 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000771 CAS Number: 112-13-0 EINECS: 203-938-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...