Ethylvanillin 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Ethoxy-4-hydroxybenzaldehyde, 3-(Ethoxy-d5)-4-hydroxybenzaldehyde
A19478-14
A19478-22
A19478-36
CAAAAA19478-14EA
35.87
CAD
CAAAAA19478-14
CAAAA19478-22
CAAAA19478-36
Ethylvanillin 98%
Ethylvanillin
Purity: 98%
CAS No.: 121-32-4
Molecular Formula: C9H10O3
Formula Weight: 166.18
Flash Point: 127°(260°F)
Melting Point: 75-79°
Boiling Point: 285°
Beilstein Reference: 1073761
Merck Reference: 14,3859
MDL No.: MFCD00006944
Formula:
C₉H₁₀O₃ Boiling Pt: 285 °C Melting Pt: 75…79 °C Flash Pt: 127 °C(260 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006944 CAS Number: 121-32-4 EINECS: 204-464-7 Merck Index: 14,3859 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...