2-Isopropylphenyl isothiocyanate 96%

Supplier: Thermo Scientific Chemicals
Warning

A19448-06 A19448-14
CAAAAA19448-03EA 97.08 CAD
CAAAAA19448-03 CAAAA19448-06 CAAAA19448-14
2-Isopropylphenyl isothiocyanate 96%
2-Isopropylphenyl isothiocyanate

Purity: 96% CAS No.: 36176-31-5 Molecular Formula: C10H11NS Formula Weight: 177.27 Refractive Index: 1.6050 Boiling Point: 131-132°/10mm UN No.: 2922 MDL No.: MFCD00022053

Formula: C₁₀H₁₁NS
MW: 177.27 g/mol
Boiling Pt: 131…132 °C (10 mmHg)
Density: 1.050
Storage Temperature: Ambient
MDL Number: MFCD00022053
CAS Number: 36176-31-5
UN: 2922
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR