D-(+)-Trehalose dihydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate

A19434-06 A19434-14 A19434-22
CAAAA19434-06EA 50.72 CAD
CAAAA19434-06 CAAAA19434-14 CAAAA19434-22
D-(+)-Trehalose dihydrate 99%
D(+)-Trehalose dihydrate

Purity: 99% CAS No.: 6138-23-4 Molecular Formula: C12H22O11.2H2O Formula Weight: 378.33 (342.29anhy) Physical Form: Crystalline powder Sensitivity: Hygroscopic Appearance: White Melting Point: 97-99° Beilstein Reference: 5322018 Merck Reference: 14,9580 Optical Rotation: +179° (c=2 in water) MDL No.: MFCD00071594

Formula: C₁₂H₂₂O₁₁·2H₂O
MW: 378.33 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00071594
CAS Number: 6138-23-4
EINECS: 202-739-6
Merck Index: 14,9580
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af741ee934bfe7c82395d31c21e2c19f0738d0f0&vid=59183c854dcfa367e15d46861ef4b9a7028540a9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR