2,5-Dibromotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

A19418-06 A19418-14
CAAAA19418-06EA 44.08 CAD
CAAAA19418-06 CAAAA19418-14
2,5-Dibromotoluene 98%
2,5-Dibromotoluene

Purity: 98% CAS No.: 615-59-8 Molecular Formula: C7H6Br2 Formula Weight: 249.94 Flash Point: 110°(230°F) Refractive Index: 1.6020 Odor: Aromatic Melting Point: 5-6° Boiling Point: 106-108°/10mm Beilstein Reference: 1859123 MDL No.: MFCD00000090

Formula: C₇H₆Br₂
Boiling Pt: 106…108 °C/10 mm
Melting Pt: 5…6 °C
Density: 1,813
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000090
CAS Number: 615-59-8
EINECS: 210-437-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR