3-Bromo-4-hydroxybenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19400-14
A19400-06
CAAAAA19400-14EA
241.74
CAD
CAAAAA19400-14
CAAAAA19400-22
CAAAA19400-06
3-Bromo-4-hydroxybenzonitrile 98%
3-Bromo-4-hydroxybenzonitrile
Purity: 98%
CAS No.: 2315-86-8
Molecular Formula: C7H4BrNO
Formula Weight: 198.02
Melting Point: 156-161°
Beilstein Reference: 2207020
UN No.: 3439
MDL No.: MFCD00143096
Formula:
C₇H₄BrNO Boiling Pt: 406 °C (1013 hPa) Melting Pt: 156…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143096 CAS Number: 2315-86-8 EINECS: 219-022-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...