5-Bromo-o-anisaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

A19396-14 A19396-22
CAAAAA19396-36EA 867.67 CAD
CAAAAA19396-36 CAAAA19396-14 CAAAA19396-22
5-Bromo-o-anisaldehyde 98+%
5-Bromo-o-anisaldehyde

Purity: 98+% CAS No.: 25016-01-7 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 114-118° Beilstein Reference: 508840

Formula: C₈H₇BrO₂
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00003347
CAS Number: 25016-01-7
EINECS: 246-564-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR