6-Methylcoumarin 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
6-Methyl-2-benzopyrone, 6-Methyl-2H-chromen-2-one
A19382-22
A19382-18
CAAAAA19382-22EA
109.65
CAD
CAAAAA19382-22
CAAAA19382-18
CAAAA19382-30
6-Methylcoumarin 99%
6-Methylcoumarin
Purity: 99%
CAS No.: 92-48-8
Molecular Formula: C10H8O2
Formula Weight: 160.17
Melting Point: 74-77°
Boiling Point: 303°/725mm
MDL No.: MFCD00006875
Formula:
C₁₀H₈O₂ MW: 160.17 g/mol Boiling Pt: 303 Melting Pt: 74…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006875 CAS Number: 92-48-8 EINECS: 202-158-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...