2,3,4,5,6-Pentafluoroaniline 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aminopentafluorobenzene, Pentafluoroaniline

A19348-06 A19348-14 A19348-22
CAAAA19348-06EA 54.67 CAD
CAAAA19348-06 CAAAA19348-14 CAAAA19348-22
2,3,4,5,6-Pentafluoroaniline 98+%
2,3,4,5,6-Pentafluoroaniline

Purity: 98+% CAS No.: 771-60-8 Molecular Formula: C6H2F5N Formula Weight: 183.08 Flash Point: 73°(163°F) Melting Point: 34-37° Boiling Point: 156-157° Beilstein Reference: 1819387 MDL No.: MFCD00007643

Formula: C₆H₂F₅N
MW: 183.08 g/mol
Boiling Pt: 156…157 °C
Melting Pt: 34…37 °C
Density: 1.744
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007643
CAS Number: 771-60-8
EINECS: 212-234-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR