2-Chloro-1-iodo-4-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloro-4-iodonitrobenzene

A19340-14 A19340-03 A19340-06
CAAAAA19340-14EA 831.08 CAD
CAAAAA19340-14 CAAAA19340-03 CAAAA19340-06
2-Chloro-1-iodo-4-nitrobenzene 98%
2-Chloro-1-iodo-4-nitrobenzene

Purity: 98% CAS No.: 41252-96-4 Molecular Formula: C6H3ClINO2 Formula Weight: 283.45 Melting Point: 97-101° Beilstein Reference: 2502411

Formula: C₆H₃ClINO₂
MW: 283.45 g/mol
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00024589
CAS Number: 41252-96-4
EINECS: 255-278-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR