Sodium toluene-4-sulfonate 90+%

Supplier: Thermo Scientific Chemicals
Warning

A19332-18 A19332-30
CAAAA19332-18EA 57.04 CAD
CAAAA19332-18 CAAAA19332-30
Sodium toluene-4-sulfonate 90+%
Sodium toluene-4-sulphonate

Purity: 90+% CAS No.: 657-84-1 Molecular Formula: C7H7NaO3S Formula Weight: 194.19(anhy) Merck Reference: 14,9533 MDL No.: MFCD00798566

Formula: C₇H₇NaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00798566
CAS Number: 657-84-1
EINECS: 211-522-5
Merck Index: 14,9533

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR