α'-Bromo-α,α,α-trifluoro-m-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(Trifluoromethyl)benzyl bromide, 1-(Bromomethyl)-3-(trifluoromethyl)benzene, 3-(Bromomethyl)benzotrifluoride
A19330-06
A19330-14
CAAAA19330-06EA
40.61
CAD
CAAAA19330-06
CAAAA19330-14
α'-Bromo-α,α,α-trifluoro-m-xylene 98%
α'-Bromo-α,α,α-trifluoro-m-xylene
Purity: 98%
CAS No.: 402-23-3
Molecular Formula: C8H6BrF3
Formula Weight: 239.04
Flash Point: 87°(188°F)
Refractive Index: 1.4920
Odor: Pungent
Boiling Point: 67-69°/4mm
Beilstein Reference: 743243
UN No.: 3265
MDL No.: MFCD00000395
Formula:
C₈H₆BrF₃ MW: 239.03 g/mol Boiling Pt: 67…69 °C/4 mm Density: 1.565 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000395 CAS Number: 402-23-3 EINECS: 206-938-9 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...