(±)-δ-Hexalactone 98%
Supplier: Thermo Scientific Chemicals
A19274-14
A19274-22
CAAAA19274-14EA
85.32
CAD
CAAAA19274-14
CAAAA19274-22
(±)-δ-Hexalactone 98%
(±)-δ-Hexalactone
Purity: 98%
CAS No.: 823-22-3
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 105°(221°F)
Refractive Index: 1.4510
Melting Point: 18°
Boiling Point: 231°
Beilstein Reference: 80501
MDL No.: MFCD00083574
Formula:
C₆H₁₀O₂ Boiling Pt: 231 °C Melting Pt: 18 °C Density: 1.040 Flash Pt: 105 °C(221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00083574 CAS Number: 823-22-3 EINECS: 212-511-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...