3,4,5-Trihydroxybenzaldehyde, hydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

A19270-03 A19270-06
CAAAA19270-03EA 175.38 CAD
CAAAA19270-03 CAAAA19270-06
3,4,5-Trihydroxybenzaldehyde, hydrate 97%
3,4,5-Trihydroxybenzaldehyde

3,4,5-trihydroxybenzaldehyde from Geum japonicum has dual inhibitory effect on matrix metalloproteinase 9; inhibition of gelatinoytic activity as well as MMP-9 expression in TNF-α induced HASMC.


Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Formula: C₇H₆O₄
Melting Pt: ∼ 212 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003371
CAS Number: 13677-79-7
EINECS: 237-168-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR