5-Bromo-2-thenoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromothiophene-2-carboxylic acid, 5-Bromo-2-thiophenecarboxylic acid

A19242-14 A19242-03 A19242-06
CAAAAA19242-14EA 532.46 CAD
CAAAAA19242-14 CAAAA19242-03 CAAAA19242-06
5-Bromo-2-thenoic acid 98%
5-Bromo-2-thenoic acid

Purity: 98% CAS No.: 7311-63-9 Molecular Formula: C5H3BrO2S Formula Weight: 207.05 Melting Point: 139-143° Beilstein Reference: 118364

Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
Melting Pt: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00079725
CAS Number: 7311-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR