p-Cymene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Isopropyl-4-methylbenzene, 4-Methylcumene, 4-Isopropyltoluene
A19226-0F
A19226-AP
CAAAA19226-0FEA
150.89
CAD
CAAAA19226-0F
CAAAA19226-AP
p-Cymene 97%
p-Cymene
Purity: 97%
CAS No.: 99-87-6
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 47°(116°F)
Refractive Index: 1.4900
Odor: Aromatic
Melting Point: -68°
Boiling Point: 176-178°
Beilstein Reference: 1903377
Merck Reference: 14,2763
UN No.: 2046
MDL No.: MFCD00008893
Formula:
C₁₀H₁₄ MW: 134.22 g/mol Boiling Pt: 176 Melting Pt: -67.9 Density: 0.860 Flash Pt: 47 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008893 CAS Number: 99-87-6 EINECS: 202-796-7 UN: 2046 ADR: 3 III Merck Index: 14,2763 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...