5-tert-Butyl-m-xylene 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
1-tert-Butyl-3,5-dimethylbenzene, 1-(tert-Butyl)-3,5-dimethylbenzene
A19220-22
A19220-36
CAAAA19220-22EA
59.57
CAD
CAAAA19220-22
CAAAA19220-36
5-tert-Butyl-m-xylene 98%
5-tert-Butyl-m-xylene
Purity: 98%
CAS No.: 98-19-1
Molecular Formula: C12H18
Formula Weight: 162.27
Flash Point: 84°(183°F)
Refractive Index: 1.4980
Melting Point: -22°
Boiling Point: 207-209°
Beilstein Reference: 1853314
MDL No.: MFCD00008832
Formula:
C₁₂H₁₈ MW: 162.27 g/mol Boiling Pt: 205 Melting Pt: -22 Density: 0.86 Flash Pt: 84 °C (183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008832 CAS Number: 98-19-1 EINECS: 202-647-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...