2'-Hydroxy-5'-methoxyacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2'-Hydroxy-5'-Methoxyacetophenone, 2′-Hydroxy-5′-methoxyacetophenone

A19216-06 A19216-14
CAAAA19216-06EA 87.06 CAD
CAAAA19216-06 CAAAA19216-14 CAAAA19216-22
2'-Hydroxy-5'-methoxyacetophenone 99%
2'-Hydroxy-5'-methoxyacetophenone

Purity: 99% CAS No.: 705-15-7 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 48-50° Beilstein Reference: 1239364 MDL No.: MFCD00008731

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 281
Melting Pt: 48…50 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008731
CAS Number: 705-15-7
EINECS: 211-882-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR