3-Hydroxy-2-naphthoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19214-0B
A19214-30
CAAAA19214-0BEA
128.61
CAD
CAAAA19214-0B
CAAAA19214-30
3-Hydroxy-2-naphthoic acid 98%
3-Hydroxy-2-naphthoic acid
Purity: 98%
CAS No.: 92-70-6
Molecular Formula: C11H8O3
Formula Weight: 188.18
Flash Point: 220°(428°F)
Melting Point: 218-222°
Beilstein Reference: 744100
Merck Reference: 14,4835
MDL No.: MFCD00004103
Formula:
C₁₁H₈O₃ Boiling Pt: 368 °C (1013 hPa) Melting Pt: 218…222 °C Flash Pt: 220 °C(428 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004103 CAS Number: 92-70-6 EINECS: 202-180-8 Merck Index: 14,4835 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...