2-tert-Butylhydroquinone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
t-Butylhydroquinone, 2-t-Butylhydroquinone, 2-(tert-Butyl)benzene-1,4-diol(may occur to produce black solid), tert-Butylhydroquinone, TBHQ
A19206-0E
A19206-36
A19206-0B
CAAAA19206-0EEA
628.84
CAD
CAAAA19206-0E
CAAAA19206-36
CAAAA19206-0B
2-tert-Butylhydroquinone 97%
2-tert-Butylhydroquinone
Purity: 97%
CAS No.: 1948-33-0
Molecular Formula: C10H14O2
Formula Weight: 166.22
Flash Point: 171°(339°F)
Melting Point: 126-129°
Boiling Point: 295°
Beilstein Reference: 637923
UN No.: 3077
MDL No.: MFCD00002344
Formula:
C₁₀H₁₄O₂ MW: 166.22 g/mol Boiling Pt: 295 °C Melting Pt: 126…129 °C Density: 1.050 Flash Pt: 171 °C (339 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002344 CAS Number: 1948-33-0 EINECS: 217-752-2 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...