1-Bromo-2-fluoro-4-iodobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

A19190-22 A19190-06 A19190-14
CAAAAA19190-22EA 1505.74 CAD
CAAAAA19190-22 CAAAA19190-06 CAAAA19190-14
1-Bromo-2-fluoro-4-iodobenzene 97%
1-Bromo-2-fluoro-4-iodobenzene

Purity: 97% CAS No.: 136434-77-0 Molecular Formula: C6H3BrFI Formula Weight: 300.90 Flash Point: 110°(230°F) Melting Point: 34-38° Boiling Point: 80°/2.5mm Beilstein Reference: 4740251 MDL No.: MFCD00042577

Formula: C₆H₃BrFI
Boiling Pt: 80 °C/2.5 mm
Melting Pt: 34…38 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042577
CAS Number: 136434-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR