cis-2-Butene-1,4-diol 96% remainder mainly trans-isomer
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19182-0B
A19182-30
CAAAA19182-0BEA
202.24
CAD
CAAAA19182-0B
CAAAA19182-30
cis-2-Butene-1,4-diol 96% remainder mainly trans-isomer
cis-2-Butene-1,4-diol
Purity: 96%
CAS No.: 6117-80-2
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: 127°(260°F)
Refractive Index: 1.4780
Melting Point: 6-8°
Boiling Point: 131-132°/12mm
Beilstein Reference: 1679241
MDL No.: MFCD00002924
Formula:
C₄H₈O₂ Boiling Pt: 131…132 °C/12 mm Melting Pt: 6…8 °C Density: 1.070 Flash Pt: 127 °C(260 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002924 CAS Number: 6117-80-2 EINECS: 228-085-1 Merck Index: 12,02390 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...