4-Bromo-3-fluorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19142-14
A19142-09
CAAAAA19142-14EA
316
CAD
CAAAAA19142-14
CAAAA19142-09
4-Bromo-3-fluorotoluene 98%
4-Bromo-3-fluorotoluene
Purity: 98%
CAS No.: 452-74-4
Molecular Formula: C7H6BrF
Formula Weight: 189.03
Flash Point: 74°(165°F)
Refractive Index: 1.5310
Boiling Point: 187-188°
Beilstein Reference: 2500154
MDL No.: MFCD00040940
Formula:
C₇H₆BrF Boiling Pt: 187…188 °C Density: 1.494 Flash Pt: 74 °C (165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040940 CAS Number: 452-74-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...