Undec-10-enoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19122-0B
A19122-30
CAAAA19122-0BEA
199.08
CAD
CAAAA19122-0B
CAAAA19122-30
Undec-10-enoic acid 99%
Undec-10-enoic acid
Purity: 99%
CAS No.: 112-38-9
Molecular Formula: C11H20O2
Formula Weight: 184.28
Flash Point: 160°(320°F)
Refractive Index: 1.4493
Melting Point: 22-25°
Boiling Point: 126-128°/1mm
Beilstein Reference: 1762631
Merck Reference: 14,9848
MDL No.: MFCD00004442
Formula:
C₁₁H₂₀O₂ Boiling Pt: 126…128 °C/1 mm Melting Pt: 22…25 °C Density: 0.910 Flash Pt: 160 °C(320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004442 CAS Number: 112-38-9 EINECS: 203-965-8 Merck Index: 14,9848 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...