(±)-3-Nonanol 98%
Supplier: Thermo Scientific Chemicals
Warning
|
A19120-18
A19120-30
CAAAA19120-18EA
184.86
CAD
CAAAA19120-18
CAAAA19120-30
(±)-3-Nonanol 98%
(±)-3-Nonanol
Purity: 98%
CAS No.: 624-51-1
Molecular Formula: C9H20O
Formula Weight: 144.26
Flash Point: 79°(174°F)
Refractive Index: 1.4310
Boiling Point: 192-194°
Beilstein Reference: 1719453
MDL No.: MFCD00014411
Formula:
C₉H₂₀O Boiling Pt: 192…194 °C Melting Pt: –23…–22 °C Density: 0.824 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014411 CAS Number: 624-51-1 EINECS: 210-850-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...