Spermidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine

A19096-14 A19096-22 A19096-03 A19096-06
CAAAAA19096-14EA 801.06 CAD
CAAAAA19096-14 CAAAAA19096-22 CAAAA19096-03 CAAAA19096-06
Spermidine 99%
Spermidine

Binds to the polyamine modulatory site of NMDA


Purity: 99% CAS No.: 124-20-9 Molecular Formula: H2N(CH2)3NH(CH2)4NH2 Formula Weight: 145.25 Flash Point: 110°(230°F) Refractive Index: 1.4790 Melting Point: 22-25° Boiling Point: 128-130°/14mm Beilstein Reference: 1698591 Merck Reference: 14,8742 UN No.: 2735 MDL No.: MFCD00008229

Formula: C₇H₁₉N₃
Boiling Pt: 128…130 °C/14 mm
Melting Pt: 22…25 °C
Density: 0.925
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008229
CAS Number: 124-20-9
EINECS: 204-689-0
UN: 3259
ADR: 8,III
Merck Index: 14,8742

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR