2-Methyl-2-pentanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19090-14
A19090-22
CAAAA19090-14EA
219.62
CAD
CAAAA19090-14
CAAAA19090-22
2-Methyl-2-pentanol 99%
2-Methyl-2-pentanol
Purity: 99%
CAS No.: 590-36-3
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 30°(86°F)
Refractive Index: 1.4100
Odor: Alcohol-like
Melting Point: -103°
Boiling Point: 120-122°
Beilstein Reference: 1718951
UN No.: 2560
MDL No.: MFCD00004485
Formula:
C₆H₁₄O MW: 102.18 g/mol Boiling Pt: 120…122 °C Melting Pt: –103 °C Density: 0.810 Flash Pt: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004485 CAS Number: 590-36-3 EINECS: 209-681-0 UN: 2560 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...