Methyl-5-bromovalerate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 5-Bromovalerate, 5-Bromovaleric acid methyl ester

A19072-06 A19072-14 A19072-22
CAAAA19072-06EA 79.32 CAD
CAAAA19072-06 CAAAA19072-14 CAAAA19072-22
Methyl-5-bromovalerate 97%
Methyl 5-bromovalerate

Purity: 97% CAS No.: 5454-83-1 Molecular Formula: Br(CH2)4CO2CH3 Formula Weight: 195.06 Flash Point: 99°(210°F) Refractive Index: 1.4630 Boiling Point: 79-80°/4mm Beilstein Reference: 1749649 MDL No.: MFCD00000265

Formula: BrH₂C(CH₂)₃COOCH₃
MW: 195.06 g/mol
Boiling Pt: 79…80 °C/4 mm
Density: 1.363
Flash Pt: 99 °C(210 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000265
CAS Number: 5454-83-1
EINECS: 226-709-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR