2-Benzyloxyphenol 98%

Supplier: Thermo Scientific Chemicals
Warning

A19020-06 A19020-14
CAAAA19020-06EA 82.95 CAD
CAAAA19020-06 CAAAA19020-14
2-Benzyloxyphenol 98%
2-Benzyloxyphenol

Purity: 98% CAS No.: 6272-38-4 Molecular Formula: C13H12O2 Formula Weight: 200.24 Flash Point: 110°(230°F) Refractive Index: 1.5910 MDL No.: MFCD00002186

Formula: C₁₃H₁₂O₂
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 38 °C
Density: 1,143
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002186
CAS Number: 6272-38-4
EINECS: 228-461-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR