(±)-β-Citronellol 95%

Supplier: Thermo Scientific Chemicals
Warning

A19016-22 A19016-36
CAAAA19016-22EA 74.26 CAD
CAAAA19016-22 CAAAA19016-36
(±)-β-Citronellol 95%
(±)-β-Citronellol

Purity: 95% CAS No.: 106-22-9 Molecular Formula: C10H20O Formula Weight: 156.27 Flash Point: 98°(208°F) Refractive Index: 1.4575 Odor: Pleasant Melting Point: -20° Boiling Point: 221-222° Beilstein Reference: 1721507 Merck Reference: 14,2330 UN No.: 3082 MDL No.: MFCD00002935

Formula: C₁₀H₂₀O
Boiling Pt: 221…222 °C
Melting Pt: < –20 °C
Density: 0.858
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002935
CAS Number: 106-22-9
EINECS: 203-375-0
UN: 3082
ADR: 9,III
Merck Index: 14,2330

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR