cis-3-Octen-1-ol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: cis-Oct-3-en-1-ol

A19014-09 A19014-18
CAAAA19014-09EA 76.47 CAD
CAAAA19014-09 CAAAA19014-18
cis-3-Octen-1-ol 95%
cis-3-Octen-1-ol

Purity: 95% CAS No.: 20125-84-2 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 82°(179°F) Refractive Index: 1.4470 Boiling Point: 88-89°/15mm Beilstein Reference: 1720075 MDL No.: MFCD00014062

Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 88…89 °C (15 mmHg)
Density: 0.850
Flash Pt: 82 °C (179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014062
CAS Number: 20125-84-2
EINECS: 243-533-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR