Dapsone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Aminophenyl sulfone, 1-Amino-4-((4-aminophenyl)sulfonyl)benzene, 4,4'-sulfonyldianiline, 4,4'-Sulfonyldianiline, 4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Diamino diphenyl sulfone, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone, 4-[(4-Aminophenyl)sulfonyl]aniline
Purity: 98%
CAS No.: 80-08-0
Molecular Formula: C12H12N2O2S
Formula Weight: 248.30
Odor: Odorless
Melting Point: 175-178°
Beilstein Reference: 788055
Merck Reference: 14,2822
MDL No.: MFCD00007887
Formula:
C₁₂H₁₂N₂O₂S MW: 248.31 g/mol Melting Pt: 175…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007887 CAS Number: 80-08-0 EINECS: 201-248-4 Merck Index: 14,2822 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...