3'-Fluoro-4'-methoxyacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(3-Fluoro-4-methoxyphenyl)-1-ethanone

A18972-06 A18972-14
CAAAA18972-03EA 63.45 CAD
CAAAA18972-03 CAAAA18972-06 CAAAA18972-14
3'-Fluoro-4'-methoxyacetophenone 99%
3'-Fluoro-4'-methoxyacetophenone

Purity: 99% CAS No.: 455-91-4 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Melting Point: 90-94° Boiling Point: 147-148°/20mm MDL No.: MFCD00026219

Formula: C₉H₉FO₂
MW: 168.17
Boiling Pt: 147…148 °C (20 mmHg)
Melting Pt: 92
Storage Temperature: Ambient
MDL Number: MFCD00026219
CAS Number: 455-91-4
EINECS: 207-253-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR