3,4-Dibutoxy-3-cyclobutene-1,2-dione 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Squaric acid dibutylester, Dibutyl squarate, 3,4-Dibutoxycyclobut-3-ene-1,2-dione, 1,2-Dibutoxy-1-cyclobutene-3,4-dione
A18960-03
A18960-06
A18960-14
CAAAA18960-03EA
44.56
CAD
CAAAA18960-03
CAAAA18960-06
CAAAA18960-14
3,4-Dibutoxy-3-cyclobutene-1,2-dione 98%
3,4-Dibutoxy-3-cyclobutene-1,2-dione
Purity: 98%
CAS No.: 2892-62-8
Molecular Formula: C12H18O4
Formula Weight: 226.27
Flash Point:
Formula:
C₁₂H₁₈O₄ MW: 226.27 g/mol Boiling Pt: 155…158 °C (3 mmHg) Density: 1.047 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00037150 CAS Number: 2892-62-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...