cis-1,4-Dichloro-2-butene 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18956-14
A18956-22
CAAAA18956-14EA
181.7
CAD
CAAAA18956-14
CAAAA18956-22
cis-1,4-Dichloro-2-butene 95%
cis-1,4-Dichloro-2-butene
Purity: 95%
CAS No.: 1476-11-5
Molecular Formula: C4H6Cl2
Formula Weight: 125.00
Storage Temperature: 2°C to 8°C
Flash Point: 55°(132°F)
Refractive Index: 1.4900
Melting Point: -48°
Boiling Point: 151-153°
Beilstein Reference: 1719692
UN No.: 3390
MDL No.: MFCD00062950
Formula:
C₄H₆Cl₂ MW: 125 Boiling Pt: 152 Melting Pt: -48 Density: 1.188 Flash Pt: 55 °C (132 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00062950 CAS Number: 1476-11-5 EINECS: 216-021-5 UN: 3390 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...