3-Mercapto-1,2-propanediol ≥90% in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
alpha-Thioglycerol, α-Monothioglycerol, Monothioglycerol, (±)-3-Mercapto-1,2-dihydroxy propane, 1-Thioglycerol, (±)-α-Monothioglycerine, (±)-1-Thioglycerine, (±)-3-Mercapto-1,2-propanediol
A18948-18
A18948-30
CAAAA18948-18EA
91.96
CAD
CAAAA18948-18
CAAAA18948-30
3-Mercapto-1,2-propanediol ≥90% in aqueous solution
3-Mercapto-1,2-propanediol
Purity: 90+%
CAS No.: 96-27-5
Molecular Formula: C3H8O2S
Formula Weight: 108.16
Flash Point: 149°(300°F)
Refractive Index: 1.5110
Boiling Point: 117-119°/5mm
Beilstein Reference: 1732046
Merck Reference: 14,9335
MDL No.: MFCD00004879
Formula:
C₃H₈O₂S MW: 108.16 g/mol Boiling Pt: 117…119 °C/5 mm Density: 1.247 Flash Pt: 149 °C (300 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004879 CAS Number: 96-27-5 EINECS: 202-495-0 UN: 2810 ADR: 6.1,III Merck Index: 14,9335 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...