3-Ethoxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

A18940-09 A18940-88
CAAAA18940-09EA 371.3 CAD
CAAAA18940-09 CAAAA18940-88
3-Ethoxybenzaldehyde 97%
3-Ethoxybenzaldehyde

Purity: 97% CAS No.: 22924-15-8 Molecular Formula: C9H10O2 Formula Weight: 150.18 Boiling Point: 132-134/15mm MDL No.: MFCD00016606

Formula: C₉H₁₀O₂
Boiling Pt: 132…134/15 mm
Melting Pt: 21 °C
Density: 1.07
Storage Temperature: Ambient
MDL Number: MFCD00016606
CAS Number: 22924-15-8
EINECS: 245-333-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR