Bis(4-chlorophenyl) sulfone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,4'-Dichloro diphenyl sulphone, 4,4'-Sulfonylbis(chlorobenzene), 4,4'-Dichloro diphenyl sulfone, 4-Chlorophenyl sulphone, 4,4'-sulphonylbis(chlorobenzene), 4-Chlorophenyl sulfone
A18938-22
A18938-36
CAAAA18938-22EA
50.09
CAD
CAAAA18938-22
CAAAA18938-36
Bis(4-chlorophenyl) sulfone 99%
Bis(4-chlorophenyl) sulphone
Purity: 99%
CAS No.: 80-07-9
Molecular Formula: C12H8Cl2O2S
Formula Weight: 287.17
Flash Point: 233°(451°F)
Melting Point: 147-150°
Boiling Point: 249-251°/10mm
Beilstein Reference: 2052955
MDL No.: MFCD00000619
Formula:
C₁₂H₈Cl₂O₂S MW: 287.17 g/mol Boiling Pt: 249…251 °C (10 mmHg) Melting Pt: 147…150 °C Flash Pt: 233 °C (451 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000619 CAS Number: 80-07-9 EINECS: 201-247-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...