Isobutyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylpropyl isothiocyanate
A18932-06
A18932-14
CAAAA18932-06EA
56.56
CAD
CAAAA18932-06
CAAAA18932-14
CAAAA18932-22
Isobutyl isothiocyanate 97%
Isobutyl isothiocyanate
Purity: 97%
CAS No.: 591-82-2
Molecular Formula: (CH3)2CHCH2NCS
Formula Weight: 115.20
Flash Point: 24°(75°F)
Refractive Index: 1.4980
Odor: Pungent
Boiling Point: 160-162°
Beilstein Reference: 1740371
UN No.: 2924
MDL No.: MFCD00022062
Formula:
C₅H₉NS MW: 115.2 g/mol Boiling Pt: 160…162 °C Density: 0.940 Flash Pt: 24 °C (75 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00022062 CAS Number: 591-82-2 EINECS: 209-733-2 UN: 3286 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...