(3,5-Bis(trifluoromethyl)phenyl)acetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18922-03
A18922-06
CAAAA18922-03EA
40.76
CAD
CAAAA18922-03
CAAAA18922-06
(3,5-Bis(trifluoromethyl)phenyl)acetic acid 97%
(3,5-Bis(trifluoromethyl)phenyl)acetic acid
Purity: 97%
CAS No.: 85068-33-3
Molecular Formula: C10H6F6O2
Formula Weight: 272.15
Melting Point: 121-123°
Beilstein Reference: 6813447
Formula:
(C₆H₃(CF₃)₂)CH₂COOH Boiling Pt: 276 °C (1013 hPa) Melting Pt: 121…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009908 CAS Number: 85068-33-3 EINECS: 285-295-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...