4-Bromobenzhydrazide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromobenzene-1-carbohydrazide, 4-Bromobenzoic acid hydrazide, 4-Bromobenzohydrazide, 4-Bromobenzoic hydrazide
A18918-06
A18918-14
CAAAAA18918-22EA
1291.59
CAD
CAAAAA18918-22
CAAAA18918-06
CAAAA18918-14
4-Bromobenzhydrazide 98+%
4-Bromobenzhydrazide
Purity: 98+%
CAS No.: 5933-32-4
Molecular Formula: C7H7BrN2O
Formula Weight: 215.05
Melting Point: 167-169°
Beilstein Reference: 777006
Formula:
C₇H₇BrN₂O Melting Pt: 167…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007602 CAS Number: 5933-32-4 EINECS: 227-681-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...